Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATG2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575156
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. Another member of the autophagy family of proteins is ATG2B, one of two homologs of ATG2 that is essential for autophagosome formation and important for regulation of size and distribution of lipid droplets. Relatively high rates of ATG2B mutations were observed in gastric and colorectal carcinomas, suggesting that deregulating the autophagy process may contribute to cancer development.
Specifications
ATG2B | |
Polyclonal | |
Unconjugated | |
ATG2B | |
2410024A21Rik; AI047755; AI503411; ATG2 autophagy related 2 homolog B; Atg2b; autophagy related 2B; autophagy-related protein 2 homolog B; AW558123; C030004M05Rik; C14orf103; C630028L02Rik; mKIAA4067; RGD1304878 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
314415, 55102, 76559 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.3 | |
Q80XK6, Q96BY7 | |
ATG2B | |
Synthetic peptide corresponding to residues in Human ATG2B. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction