Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG3/APG3 Antibody (OTI3H2), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272140PEATT594
Description
ATG3/APG3 Monoclonal specifically detects ATG3/APG3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
ATG3/APG3 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
ATG3 | |
Full length recombinant protein of human ATG3/APG3 (NP_071933) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG2a |
Flow Cytometry | |
OTI3H2 | |
Flow Cytometry | |
APG3, APG3 autophagy 3-like (S. cerevisiae), APG3L, APG3-LIKE, ATG3 autophagy related 3 homolog (S. cerevisiae), Autophagy-related protein 3, DKFZp564M1178, EC 6.3.2.-, FLJ22125,2610016C12Rik, hApg3, MGC15201, PC3-96, Protein PC3-96, ubiquitin-like-conjugating enzyme ATG3 | |
Mouse | |
Affinity Purified | |
Autophagy, Cancer | |
64422.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction