Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG3/APG3 Rabbit anti-Human, Mouse, Rat, Clone: 5D2W0, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315800100UL
This item is not returnable.
View return policy
Description
ATG3/APG3 Monoclonal antibody specifically detects ATG3/APG3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ATG3/APG3 | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
5D2W0 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
APG3, APG3 autophagy 3-like (S. cerevisiae), APG3L, APG3-LIKE, ATG3 autophagy related 3 homolog (S. cerevisiae), Autophagy-related protein 3, DKFZp564M1178, EC 6.3.2.-, FLJ22125,2610016C12Rik, hApg3, MGC15201, PC3-96, Protein PC3-96, ubiquitin-like-conjugating enzyme ATG3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ATG3/APG3 (Q9NT62). | |
100 μg | |
Autophagy, Cancer | |
64422 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction