Learn More
Invitrogen™ ATG3 Recombinant Rabbit Monoclonal Antibody (5D2W0)

Description
Immunogen sequence: MQNVINTVKG KALEVAEYLT PVLKESKFKE TGVITPEEFV AAGDHLVHHC PTWQWATGEE LKVKAYLPTG KQFLVTKNVP CYKRCKQMEY SDELEAIIEE.
Specifications
Specifications
| Antigen | ATG3 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 5D2W0 |
| Concentration | 0.23 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ATG3 |
| Gene Accession No. | Q6AZ50, Q9CPX6, Q9NT62 |
| Gene Alias | 2610016C12Rik; APG3; Apg3l; APG3-like; Apg3p; atg3; ATG3 autophagy related 3 homolog; atg3.S; Atg3l; autophagy related 3; autophagy related 3 S homeolog; autophagy-related 3; autophagy-related protein 3; DKFZp564M1178; EGK_11288; FLJ22125; hApg3; MGC15201; PC3-96; Pig1; PIG-1; preconditioning-inducible gene 1 protein; Protein PC3-96; Ubiquitin-like-conjugating enzyme ATG3; XELAEV_18014721mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.