Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG3 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI3H2, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA503346

Description
ATG3 (APG3) is a widely expressed conjugating enzyme for APG8 lipidation, an essential step for the initiation of autophagy. Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. Autophagy is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. ATG3 functions as an E2-like enzyme during the initial stages of autophagosome formation by catalyzing the formation of the Atg8-phosphatidylethanolamine (Atg8-PE) conjugate, which is critical for autophagy.Specifications
ATG3 | |
Monoclonal | |
Unconjugated | |
ATG3 | |
APG3, APG3-LIKE, APG3L, PC3-96 | |
Mouse | |
Affinity Chromatography | |
RUO | |
171415, 478564, 64422, 67841 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3H2 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q6AZ50, Q9CPX6, Q9NT62 | |
ATG3 | |
Full length human recombit protein of human ATG3 produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction