Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG3 Mouse anti-Human, Mouse, Clone: 1377CT239.6.1.12, Abnova™

Catalog No. 89933594 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-594 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-594 Supplier Abnova Corporation Supplier No. MAB12360
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human ATG3.

Autophagy is a process of bulk degradation of cytoplasmic components by the lysosome or vacuole. Human ATG3 displays the same enzymatic characteristics in vitro as yeast Apg3, a protein-conjugating enzyme essential for autophagy (Tanida et al., 2002 [PubMed 11825910]).[supplied by OMIM]

Specifications

Antigen ATG3
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1377CT239.6.1.12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human ATG3.
Dilution Immunohistochemistry (1:25) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias APG3/APG3-LIKE/APG3L/DKFZp564M1178/FLJ22125/MGC15201/PC3-96
Gene Symbols ATG3
Host Species Mouse
Immunogen Recombinant protein corresponding to human ATG3.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64422
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.