Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144485 Supplier Invitrogen™ Supplier No. PA5144485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG32 Polyclonal Antibody for ELISA, Western Blot

Mitophagy-specific receptor that recruits the autophagic machinery to mitochondria and regulates selective degradation of mitochondria. Mitophagy contributes to regulate mitochondrial quantity and quality by eliminating the mitochondria to a basal level to fulfill cellular energy requirements and preventing excess ROS production. Recruits ATG11 to the surface of mitochondria. Promotes also autophagy-dependent peroxisome degradation (By similarity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG32
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ATG32
Gene Accession No. B3LTZ3, C7GUR3
Gene Alias ECM17
Gene Symbols ATG32
Host Species Rabbit
Immunogen Recombinant Saccharomyces cerevisiae (strain JAY291) (Baker's yeast) ATG32 protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 854660
Target Species Yeast
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.