Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG4A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | ATG4A |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ATG4A Polyclonal specifically detects ATG4A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ATG4A | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
APG4 autophagy 4 homolog A (S. cerevisiae), APG4A, ATG4 autophagy related 4 homolog A (S. cerevisiae), AUTL2, AUT-like 2 cysteine endopeptidase, AUT-like 2, cysteine endopeptidase (S. cerevisiae), autophagin 2, autophagin-2, cysteine endopeptidase, cysteine protease ATG4A, EC 3.4.22, EC 3.4.22.-, hAPG4A | |
ATG4A | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Autophagy | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
115201 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:QSPQRMNILNLDPSVALGFFCKEEKDFDNWCSLVQKEILKENLRMFELVQKHPSHWPPFVPPAKPEVTTTGAEFIDSTEQLEEFDLEEDFEILSV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title