Learn More
Invitrogen™ ATG4A Monoclonal Antibody (1458CT808.66.25.69)

Description
ATG4A Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | ATG4A |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1458CT808.66.25.69 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ATG4A |
| Gene Accession No. | Q8WYN0 |
| Gene Alias | AI627006; APG4 (ATG4) autophagy-related homolog A; APG4 autophagy 4 homolog A; APG4A; ATG4 autophagy related 4 homolog A; Atg4a; Atg4al; Atg4al1; AUTL2; AUT-like 2 cysteine endopeptidase; AUT-like 2, cysteine endopeptidase; autophagin 2; autophagin-2; autophagy 4 homolog A; autophagy related 4 homolog A; autophagy related 4A cysteine peptidase; autophagy related 4A, cysteine peptidase; autophagy related 4A, cysteine peptidase-like 1; autophagy-related 4A; autophagy-related cysteine endopeptidase 2; Autophagy-related protein 4 homolog A; AV169859; cysteine protease APG4A; cysteine protease ATG4A; hAPG4A; MGC107179 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.