Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111588 Supplier Invitrogen™ Supplier No. PA5111588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESVLSKYEDQ ITIFTDYLEE YPDTDELVWI LGKQHLLKTE KSKLLSDISA RLWFTYRRKF SPIGGTG Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 91%.

Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some malignant tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Transcript variants that encode distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG4A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATG4A
Gene Accession No. Q8WYN0
Gene Alias AI627006; APG4 (ATG4) autophagy-related homolog A; APG4 autophagy 4 homolog A; APG4A; ATG4 autophagy related 4 homolog A; Atg4a; Atg4al; Atg4al1; AUTL2; AUT-like 2 cysteine endopeptidase; AUT-like 2, cysteine endopeptidase; autophagin 2; autophagin-2; autophagy 4 homolog A; autophagy related 4 homolog A; autophagy related 4A cysteine peptidase; autophagy related 4A, cysteine peptidase; autophagy related 4A, cysteine peptidase-like 1; autophagy-related 4A; autophagy-related cysteine endopeptidase 2; Autophagy-related protein 4 homolog A; AV169859; cysteine protease APG4A; cysteine protease ATG4A; hAPG4A; MGC107179
Gene Symbols ATG4A
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ATG4A. Recombinant protein control fragment (Product #RP-105342).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115201
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.