Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG4B Monoclonal Antibody (6C3C5)
GREENER_CHOICE

Catalog No. PIMA538451
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538451 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538451 Supplier Invitrogen™ Supplier No. MA538451
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

Cysteine protease ATG4b is an enzyme which is coded by ATG4b gene. Autophagy is a critical cellular homeostatic process that controls the turnover of damaged organelles and protein. ATG4bis a deubiquitin-like protease that plays dual roles in the core machinery of autophagy. ATG4b, cleaves the C-terminal amino acid of ATG8 family proteins MAP1LC3, GABARAPL1, GABARAPL2 and GABARAP, to reveal a C-terminal glycine. Exposure of the glycine at the C-terminus is essential for ATG8 proteins conjugation to phosphatidylethanolamine (PE) and insertion to membranes, which is necessary for autophagy. ATG4b is mainly expressed in the skeletal muscle, brain, heart, liver and pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG4B
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 6C3C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Atg4b
Gene Accession No. Q9Y4P1
Gene Alias 2510009N07Rik; APG4 (ATG4) autophagy-related homolog B; APG4 autophagy 4 homolog B; APG4B; ATG4 autophagy related 4 homolog B; Atg4b; atg4b protein; Atg4bl; AUT2B; Aut2B1; Aut2b2; AUTL1; AUT-like 1 cysteine; AUT-like 1 cysteine endopeptidase; AUT-like 1, cysteine endopeptidase; autophagin 1; autophagin-1; Autophagin-2B; autophagy related 4 homolog B; autophagy related 4B cysteine peptidase; autophagy related 4B, cysteine peptidase; autophagy-related 4B; autophagy-related cysteine endopeptidase 1; Autophagy-related cysteine endopeptidase 2B; autophagy-related protein 4 homolog B; AW048066; bAut2B; Cysteine protease ATG4B; cysteine protease involved in autophagy APG4-B; hAPG4B; KIAA0943
Gene Symbols Atg4b
Host Species Mouse
Immunogen Purified recombinant fragment of human ATG4B (aa: 1-221) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23192
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.