Learn More
ATG4B Mouse anti-Human, Clone: OTI1A3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810189

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cysteine protease ATG4b is an enzyme which is coded by ATG4b gene. Autophagy is a critical cellular homeostatic process that controls the turnover of damaged organelles and protein. ATG4bis a deubiquitin-like protease that plays dual roles in the core machinery of autophagy. ATG4b, cleaves the C-terminal amino acid of ATG8 family proteins MAP1LC3, GABARAPL1, GABARAPL2 and GABARAP, to reveal a C-terminal glycine. Exposure of the glycine at the C-terminus is essential for ATG8 proteins conjugation to phosphatidylethanolamine (PE) and insertion to membranes, which is necessary for autophagy. ATG4b is mainly expressed in the skeletal muscle, brain, heart, liver and pancreas.Specifications
ATG4B | |
Monoclonal | |
Unconjugated | |
ATG4B | |
APG4B, AUTL1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23192 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1A3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y4P1 | |
ATG4B | |
Full length human recombit protein of human ATG4B produced in HEK293T Cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.