Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG4B Rabbit anti-Human, Mouse, Rat, Clone: 9H6M6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | ATG4B |
---|---|
Clone | 9H6M6 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
ATG4B Monoclonal antibody specifically detects ATG4B in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
ATG4B | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Autophagy, Cancer | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
23192 | |
IgG | |
Affinity purified |
9H6M6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
APG4 autophagy 4 homolog B, APG4 autophagy 4 homolog B (S. cerevisiae), APG4B, ATG4 autophagy related 4 homolog B (S. cerevisiae), AUTL1MGC1353, AUT-like 1 cysteine endopeptidase, Autophagin-1, Autophagy-related cysteine endopeptidase 1, Autophagy-related protein 4 homolog B, cysteine protease ATG4B, DKFZp586D1822, EC 3.4.22, EC 3.4.22.-, hAPG4B, KIAA0943 | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ATG4B (Q9Y4P1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title