Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501730006 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0006 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0006 Supplier Proteintech Group Inc Supplier No. 151311AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

QKI belongs to a family of RNA-binding proteins that have an HNRNPK. QKI proteins regulate RNA splicing, export of target RNAs from the nucleus, translation of proteins, and RNA stability.

Specifications

Antigen ATG4B
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATG4B
Gene Accession No. Q8BGE6, Q9Y4P1
Gene Alias APG4B, ATG4B, AUTL1, Autophagin 1, Cysteine protease ATG4B, hAPG4B, KIAA0943
Gene Symbols Atg4b
Host Species Rabbit
Immunogen ATG4B Fusion Protein Ag7261
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23192, 316640, 66615
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.