Learn More
ATG4B Recombinant Rabbit Monoclonal Antibody (1HC6LC10)
Description
ATG4B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | ATG4B |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 1HC6LC10 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Atg4b |
| Gene Accession No. | Q8BGE6, Q9Y4P1 |
| Gene Alias | 2510009N07Rik; APG4 (ATG4) autophagy-related homolog B; APG4 autophagy 4 homolog B; APG4B; ATG4 autophagy related 4 homolog B; Atg4b; atg4b protein; Atg4bl; AUT2B; Aut2B1; Aut2b2; AUTL1; AUT-like 1 cysteine; AUT-like 1 cysteine endopeptidase; AUT-like 1, cysteine endopeptidase; autophagin 1; autophagin-1; Autophagin-2B; autophagy related 4 homolog B; autophagy related 4B cysteine peptidase; autophagy related 4B, cysteine peptidase; autophagy-related 4B; autophagy-related cysteine endopeptidase 1; Autophagy-related cysteine endopeptidase 2B; autophagy-related protein 4 homolog B; AW048066; bAut2B; Cysteine protease ATG4B; cysteine protease involved in autophagy APG4-B; hAPG4B; KIAA0943 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.