Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG4C Antibody (2E10H7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261691
Description
ATG4C Monoclonal specifically detects ATG4C in Human samples. It is validated for Western Blot, ELISA.Specifications
ATG4C | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
APG4 autophagy 4 homolog C, APG4 autophagy 4 homolog C (S. cerevisiae), APG4C, ATG4 autophagy related 4 homolog C (S. cerevisiae), AUT (S. cerevisiae)-like 1, cysteine endopeptidase; AUT-like 1, cysteineendopeptidase (S. cerevisiae), AUTL1, AUTL3APG4-C, AUT-like 1, cysteine endopeptidase, AUT-like 3 cysteine endopeptidase, Autophagin-3, Autophagy-related cysteine endopeptidase 3, Autophagy-related protein 4 homolog C, cysteine protease ATG4C, EC 3.4.22, EC 3.4.22.-, FLJ14867 | |
Mouse | |
52.5 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
2E10H7 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
ATG4C | |
Purified recombinant fragment of human ATG4C (AA: 321-458) expressed in E. Coli. | |
Protein G purified | |
Autophagy, Cancer | |
84938 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction