Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG4C Monoclonal Antibody (2E10H7)
GREENER_CHOICE

Catalog No. PIMA531640
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531640 Supplier Invitrogen™ Supplier No. MA531640
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31640 has been tested in indirect ELISA.

ATG4C is involved in autophagy, the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some malignant tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Alternate transcriptional splice variants, encoding the same protein, have been characterized. Cysteine protease required for autophagy, which cleaves the C-terminal part of either MAP1LC3, GABARAPL2 or GABARAP, allowing the liberation of form I. A subpopulation of form I is subsequently converted to a smaller form (form II). Form II, with a revealed C-terminal glycine, is considered to be the phosphatidylethanolamine (PE)-conjugated form, and has the capacity for the binding to autophagosomes. Highly expressed in skeletal muscle, heart, liver and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG4C
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E10H7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene ATG4C
Gene Accession No. Q96DT6
Gene Alias APG4 (ATG4) autophagy-related homolog C; APG4 autophagy 4 homolog C; APG4C; APG4-C; ATG4 autophagy related 4 homolog C; ATG4C; Atg4cl; AUTL1; AUTL3; AUT-like 1, cysteine endopeptidase; AUT-like 3 cysteine endopeptidase; autophagin 3; autophagin-3; autophagy related 4C cysteine peptidase; autophagy related 4C, cysteine peptidase; autophagy-related 4C; autophagy-related cysteine endopeptidase 3; Autophagy-related protein 4 homolog C; cysteine peptidase; cysteine protease ATG4C; cysteine protease involved in autophagy; FLJ14867
Gene Symbols ATG4C
Host Species Mouse
Immunogen Purified recombinant fragment of human ATG4C (AA: 321-458) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84938
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.