Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG4C Mouse anti-Human, Clone:2E10H7, Abnova™

Catalog No. 89334417 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-417 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-417 Supplier Abnova Corporation Supplier No. MAB17927
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human ATG4C.

Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some malignant tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Alternate transcriptional splice variants, encoding the same protein, have been characterized. [provided by RefSeq]

Specifications

Antigen ATG4C
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E10H7
Conjugate Unconjugated
Description ATG4 autophagy related 4 homolog C (S. cerevisiae)
Dilution ELISA (1:10000)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene ATG4C
Gene Alias APG4-C, APG4C, AUTL1, AUTL3, FLJ14867
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 321-458 of human ATG4C.
Quantity 100 μg
Gene ID (Entrez) 84938
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.