Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATG4D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598576
Description
ATG4D Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Atg4D (autophagy-related gene 4D), also known as APG4D or AUTL4, is a 474 amino acid protein that localizes to the cytoplasm and belongs to the C-54 family of cysteine proteases. Expressed predominately in skeletal muscle, but also present in testis, Atg4D functions as a cysteine protease that is required for autophagy and functions to specifically cleave the C-terminal region of target proteins, thereby allowing the target proteins to bind to autophagosomes. The enzymatic activity of Atg4D is inhibited by Nethylmaleimide, a thiol reactive compound that is capable of modifying cystine residues in proteins and peptides.
Specifications
ATG4D | |
Polyclonal | |
Unconjugated | |
Atg4d | |
9830134P12Rik; APG4 (ATG4) autophagy-related homolog D; APG4 autophagy 4 homolog D; APG4D; APG4-D; ATG4 autophagy related 4 homolog D; Atg4d; Atg4dl; Autl4; AUT-like 4 cysteine endopeptidase; AUT-like 4, cysteine endopeptidase; Autophagin-4; autophagy related 4D cysteine peptidase; autophagy related 4D, cysteine peptidase; autophagy-related 4D; autophagy-related cysteine endopeptidase 4; Autophagy-related protein 4 homolog D; cysteine protease ATG4D; Cysteine protease ATG4D, mitochondrial; cysteine protease involved in autophagy; cysteine protease involved in autophagy APG4-D | |
Rabbit | |
Protein G | |
RUO | |
235040, 84971 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
3.73 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q86TL0, Q8BGV9 | |
Atg4d | |
Recombinant Human Cysteine protease ATG4D protein (14-43AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction