Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG4D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596828 Supplier Invitrogen™ Supplier No. PA596828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVTRLVVYVS QDCTVYKADV ARLVARPDPT AEWKSVVILV PVRLGGETLN PVYVPCVKEL LRCELCLGIM GGKPRHSLYF IGYQDDFLLY LDPHYCQPTV DVSQADFPLE SFHCTSPRKM AFAKMDPSCT VGFYAGDRKE FETLCSELTR VLSSSSATER YPMFTLAEGH AQDHSLDDLC SQLAQPTLRL PRTGRLLRAK RPSSEDFVFL Positive Samples: 22Rv1, NCI-H460, Mouse heart; Cellular Location: Cytoplasm, Mitochondrion matrix.

Atg4D (autophagy-related gene 4D), also known as APG4D or AUTL4, is a 474 amino acid protein that localizes to the cytoplasm and belongs to the C-54 family of cysteine proteases. Expressed predominately in skeletal muscle, but also present in testis, Atg4D functions as a cysteine protease that is required for autophagy and functions to specifically cleave the C-terminal region of target proteins, thereby allowing the target proteins to bind to autophagosomes. The enzymatic activity of Atg4D is inhibited by Nethylmaleimide, a thiol reactive compound that is capable of modifying cystine residues in proteins and peptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG4D
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Atg4d
Gene Accession No. Q86TL0, Q8BGV9
Gene Alias 9830134P12Rik; APG4 (ATG4) autophagy-related homolog D; APG4 autophagy 4 homolog D; APG4D; APG4-D; ATG4 autophagy related 4 homolog D; Atg4d; Atg4dl; Autl4; AUT-like 4 cysteine endopeptidase; AUT-like 4, cysteine endopeptidase; Autophagin-4; autophagy related 4D cysteine peptidase; autophagy related 4D, cysteine peptidase; autophagy-related 4D; autophagy-related cysteine endopeptidase 4; Autophagy-related protein 4 homolog D; cysteine protease ATG4D; Cysteine protease ATG4D, mitochondrial; cysteine protease involved in autophagy; cysteine protease involved in autophagy APG4-D
Gene Symbols Atg4d
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 265-474 of human ATG4D (NP_1162743).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235040, 686505, 84971
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.