Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG5 Antibody (603813) [Janelia Fluor™ 635], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5294JF635
Description
ATG5 Monoclonal antibody specifically detects ATG5 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Simple WesternSpecifications
| ATG5 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| APG5, APG5 (Autophagy 5, S. Cerevisiae)-Like, APG5 autophagy 5-like (S. cerevisiae), APG5L, APG5-LIKE, ASP, ASPAPG5-LIKE, ATG5 Autophagy Related 5 Homolog, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5, Autophagy Related 5, HAPG5, SCAR25 | |
| E. coli-derived recombinant human ATG5, Asn99-Thr193, Accession # Q9H1Y0 | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry, Western Blot, Simple Western | |
| 603813 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Ubiquitin Proteasome Pathway | |
| 9474 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction