Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG5 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224389AF594
Description
ATG5 Polyclonal specifically detects ATG5 in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ATG5 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
APG5 autophagy 5-like (S. cerevisiae), ASPAPG5-LIKE, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ATG5 | |
A portion of amino acid 1-50 of human APG5L was used as the immunogen. | |
0.1 mL | |
Apoptosis, Autophagy, Cancer, Cell Biology, Ubiquitin Proteasome Pathway | |
Human, Mouse, Rat, Pig, Bovine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction