Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG5 Antibody (ATG5/2492) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308433
Description
ATG5 Monoclonal specifically detects ATG5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.Specifications
ATG5 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 ug/mL, ELISA : 1-5 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL | |
APG5, APG5 (Autophagy 5, S. Cerevisiae)-Like, APG5 autophagy 5-like (S. cerevisiae), APG5L, APG5-LIKE, ASP, ASPAPG5-LIKE, ATG5 Autophagy Related 5 Homolog, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5, Autophagy Related 5, HAPG5, SCAR25 | |
Recombinant fragment of human ATG5 protein (around aa 1-119) (exact sequence is proprietary) | |
100 μg | |
Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Ubiquitin Proteasome Pathway | |
9474 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
ATG5/2492 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction