Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG5 Monoclonal Antibody (8E8G6)
GREENER_CHOICE

Catalog No. PIMA538452
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538452 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538452 Supplier Thermo Scientific Supplier No. MA538452
Add to Cart
Add to Cart

ATG5 Monoclonal Antibody (8E8G6)

ATG5 Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG5
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8E8G6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Atg5
Gene Accession No. Q9H1Y0
Gene Alias 2010107M05Rik; 3110067M24Rik; APG 5; APG5; APG5 autophagy 5 like (APG5L); APG5 autophagy 5-like protein; APG5 autophagy 5-like protein, isoform 1; APG5L; APG5-LIKE; apoptosis-specific protein; Apoptosis-specific protein (ASP); ASP; atg5; Atg-5; ATG5 autophagy related 5 homolog; ATG5 autophagy related 5 homolog (S. cerevisiae); atg5.L; Atg5l; Atg5-PA; autophagy 5-like protein; autophagy protein 5; autophagy related 5; autophagy related 5 L homeolog; autophagy related 5-like protein; Autophagy-related 5; autophagy-specific gene 5; AW319544; C88337; CG1643; CG1643-PA; DmAtg5; Dmel\CG1643; Dmel_CG1643; hAPG5; hypothetical protein LOC532686; LD34980p; Paddy; XELAEV_18027227mg; zgc:100934
Gene Symbols Atg5
Host Species Mouse
Immunogen Synthesized peptide of human ATG5 (aa: MTDDKDVLRDVWFGRIc) (aa 1-16).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9474
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.