Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG5 Recombinant Rabbit Monoclonal Antibody (8L7X6)
GREENER_CHOICE

Catalog No. PIMA535339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535339 Supplier Invitrogen™ Supplier No. MA535339
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MTDDKDVLRD VWFGRIPTCF TLYQDEITER EAEPYYLLLP RVSYLTLVTD KVKKHFQKVM RQEDISEIWF EYEGTPLKWH YPIGLLFDLL ASSSALPWNI.

ATG5 (Autophagy Related 5) is an important element for autophagy and may play an important role in the apoptotic process. ATG5 is also involved in other cellular processes that include mitochondrial quality control after oxidative damage, negative regulation of the innate anti-viral immune response, lymphocyte development and proliferation, MHC II antigen presentation, and adipocyte differentiation. Following conjugation to ATG12, the conjugate participates in the formation of autophagosome. ATG5 contributes to autophagic cell death by interacting with Fas-associated protein with death domain (FADD). The ATG5-ATG12 conjugate forms a cup-shaped isolation membrane that then detaches from the membrane immediately before or after autophagosome formation is completed. APG5 may play a role in the apoptotic process, possibly within the modified cytoskeleton. Further, APG5 expression is a relatively late event in the apoptotic process, occurring downstream of caspase activity. The APG5-APG12 conjugate also associates with innate immune response proteins such as RIG-I and VISA (also known as IPS-1), inhibiting type I interferon production and permitting viral replication in host cells. Diseases associated with ATG5 dysfunction include spinocerebellar ataxia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG5
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 8L7X6
Concentration 0.70 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.09% sodium azide; pH 7.3
Gene Atg5
Gene Accession No. Q3MQ06, Q99J83, Q9H1Y0
Gene Alias 2010107M05Rik; 3110067M24Rik; APG 5; APG5; APG5 autophagy 5 like (APG5L); APG5 autophagy 5-like protein; APG5 autophagy 5-like protein, isoform 1; APG5L; APG5-LIKE; apoptosis-specific protein; Apoptosis-specific protein (ASP); ASP; atg5; Atg-5; ATG5 autophagy related 5 homolog; ATG5 autophagy related 5 homolog (S. cerevisiae); atg5.L; Atg5l; Atg5-PA; autophagy 5-like protein; autophagy protein 5; autophagy related 5; autophagy related 5 L homeolog; autophagy related 5-like protein; Autophagy-related 5; autophagy-specific gene 5; AW319544; C88337; CG1643; CG1643-PA; DmAtg5; Dmel\CG1643; Dmel_CG1643; hAPG5; hypothetical protein LOC532686; LD34980p; Paddy; XELAEV_18027227mg; zgc:100934
Gene Symbols Atg5
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11793, 365601, 9474
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.