Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG5 Mouse anti-Human, Mouse, Clone: 2G3, Abnova™

Catalog No. 89933573 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-573 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-573 Supplier Abnova Corporation Supplier No. H00009474M07
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ATG5.

Sequence: PAEENGFRYIPFRIYQTTTERPFIQKLFRPVAADGQLHTLGDLLKEVCPSAIDPEDGEKKNQVMIHGIEPMLETPLQWLSEHLSYPDNFLHISIIPQPTD

Specifications

Antigen ATG5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ATG5.
Formulation PBS with no preservative; pH 7.4
Gene ATG5
Gene Accession No. BC002699
Gene Alias APG5/APG5-LIKE/APG5L/ASP/hAPG5
Gene Symbols ATG5
Host Species Mouse
Immunogen ATG5 (AAH02699,176 a.a. ∽ 275 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9474
Test Specificity Antibody Reactive Against Recombinant Protein.
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.