Learn More
Invitrogen™ ATG5 Polyclonal Antibody, DyLight™ 550

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522858
Description
In Western Blot, a band is seen ~56 kDa representing the ATG5-ATG12 complex, the molecular weight of human ATG5 is ~33 kDa. Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended. Suggested positive control: mouse wildtype ES cell lysate.
ATG5 (Autophagy Related 5) is an important element for autophagy and may play an important role in the apoptotic process. ATG5 is also involved in other cellular processes that include mitochondrial quality control after oxidative damage, negative regulation of the innate anti-viral immune response, lymphocyte development and proliferation, MHC II antigen presentation, and adipocyte differentiation. Following conjugation to ATG12, the conjugate participates in the formation of autophagosome. ATG5 contributes to autophagic cell death by interacting with Fas-associated protein with death domain (FADD). The ATG5-ATG12 conjugate forms a cup-shaped isolation membrane that then detaches from the membrane immediately before or after autophagosome formation is completed. APG5 may play a role in the apoptotic process, possibly within the modified cytoskeleton. Further, APG5 expression is a relatively late event in the apoptotic process, occurring downstream of caspase activity. The APG5-APG12 conjugate also associates with innate immune response proteins such as RIG-I and VISA (also known as IPS-1), inhibiting type I interferon production and permitting viral replication in host cells. Diseases associated with ATG5 dysfunction include spinocerebellar ataxia.
Specifications
| ATG5 | |
| Polyclonal | |
| DyLight 550 | |
| Atg5 | |
| 2010107M05Rik; 3110067M24Rik; APG 5; APG5; APG5 autophagy 5 like (APG5L); APG5 autophagy 5-like protein; APG5 autophagy 5-like protein, isoform 1; APG5L; APG5-LIKE; apoptosis-specific protein; Apoptosis-specific protein (ASP); ASP; atg5; Atg-5; ATG5 autophagy related 5 homolog; ATG5 autophagy related 5 homolog (S. cerevisiae); atg5.L; Atg5l; Atg5-PA; autophagy 5-like protein; autophagy protein 5; autophagy related 5; autophagy related 5 L homeolog; autophagy related 5-like protein; Autophagy-related 5; autophagy-specific gene 5; AW319544; C88337; CG1643; CG1643-PA; DmAtg5; Dmel\CG1643; Dmel_CG1643; hAPG5; hypothetical protein LOC532686; LD34980p; Paddy; XELAEV_18027227mg; zgc:100934 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100735170, 100739102, 11793, 31666, 365601, 443756, 494180, 532686, 641362, 9474 | |
| 4°C, store in dark | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot, Proximity Ligation Assay, Immunocytochemistry | |
| 0.82 mg/mL | |
| 50mM sodium borate with 0.05% sodium azide | |
| Q3MQ04, Q3MQ06, Q3MQ24, Q99J83, Q9H1Y0, Q9W3R7 | |
| Atg5, atg5.L | |
| Synthetic peptide of an N-terminal region of the human ATG5 protein (within residues 1-50). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Bovine, Pig, Fish, Drosophila, Guinea Pig, Reptile, Frog, Frog | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.