Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG7 Monoclonal Antibody (29CT10.3.12)
GREENER_CHOICE

Catalog No. PIMA537466
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537466 Supplier Invitrogen™ Supplier No. MA537466
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATG7 Monoclonal Antibody for Western Blot

This gene was identified based on homology to Pichia pastoris GSA7 and Saccharomyces cerevisiae APG7. In the yeast, the protein appears to be required for fusion of peroxisomal and vacuolar membranes. The protein shows homology to the ATP-binding and catalytic sites of the E1 ubiquitin activating enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG7
Applications Western Blot
Classification Monoclonal
Clone 29CT10.3.12
Concentration Conc. not determined
Conjugate Unconjugated
Formulation ascites with 0.09% sodium azide; pH 7.4
Gene ATG7
Gene Accession No. O95352
Gene Alias 1810013K23Rik; Agp7; APG7L; APG7-LIKE; ATG12-activating enzyme E1 ATG7; Atg7; ATG7 autophagy related 7 homolog; Atg7l; autophagy related 7; autophagy related 7 homolog; autophagy-related 7; autophagy-related protein 7; DKFZp434N0735; Gm21553; GSA7; hAGP7; ubiquitin-activating enzyme E1-like protein; Ubiquitin-like modifier-activating enzyme ATG7
Gene Symbols ATG7
Host Species Mouse
Immunogen purified His-tagged APG7 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10533
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.