Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG8 Rabbit anti-Drosophila, KHL, Polyclonal, MilliporeSigma™

Catalog No. ABC974I10MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC974I10MI 100 μg
ABC974I25MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABC974I10MI Supplier MilliporeSigma Supplier No. ABC974I100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG8 Polyclonal antibody specifically detects ATG8 in Drosophila samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG8
Applications Western Blot
Classification Polyclonal
Conjugate KLH
Gene Accession No. NP_727447, NP_650649
Gene Alias Autophagy-related 8a;isoform A and Autophagy-related 8b;isoform B
Gene Symbols ATG8a, ATG8b
Host Species Rabbit
Immunogen KLH-conjugated linear peptides corresponding to 15 amino acids from the N-terminal region of Drosophila melanogaster ATG8a and ATG8b.
Molecular Weight of Antigen ∼14 kDa observed; 14.39 and 14.46 kDa calculated for ATG8a and ATG8b, respectively. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects ATG8a and ATG8b in Drosophila melanogaster. It targets an epitope within 15 amino acids from the N-terminal region.
Target Species Drosophila
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.