Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG9A Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11056893AF594
Description
ATG9A Polyclonal specifically detects ATG9A in Human, Mouse, Rat, Alligator, Bovine, Chicken, Drosophila, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| ATG9A | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| APG9 autophagy 9-like 1, APG9 autophagy 9-like 1 (S. cerevisiae), APG9L1APG9-like 1, ATG9 autophagy related 9 homolog A (S. cerevisiae), autophagy 9-like 1 protein, autophagy-related protein 9A, FLJ22169, mATG9, MGD3208 | |
| Rabbit | |
| Affinity Purified | |
| Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Phospho Specific, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
| 79065.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ATG9A | |
| A synthetic peptide made to a region within the C-terminus of human ATG9A (between residues 750-839). [Swiss-Prot: Q7Z3C6]. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Alligator, Bovine, Chicken, Drosophila, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction