Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG9A Antibody (Atg9 14F2 8B1), DyLight 405, Novus Biologicals™

Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP250109V
Description
ATG9A Monoclonal specifically detects ATG9A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
ATG9A | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ATG9A | |
Conserved C-terminal synthetic peptide of Human ATG9A sequence | |
0.1 mL | |
Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
79065 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
Atg9 14F2 8B1 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
APG9 autophagy 9-like 1, APG9 autophagy 9-like 1 (S. cerevisiae), APG9L1APG9-like 1, ATG9 autophagy related 9 homolog A (S. cerevisiae), autophagy 9-like 1 protein, autophagy-related protein 9A, FLJ22169, mATG9, MGD3208 | |
Hamster | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction