Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG9A Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 22 publications

$206.50 - $499.50

Specifications

Antigen ATG9A
Concentration 1.0 mg/ml
Dilution Western Blot 2 ug/ml, Flow Cytometry, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24705551), Immunoprecipitation reported in scientific literature (PMID 27316455), Immunohistochemistry-Paraffin 1:100-1:250, Immunohistochemistry-Frozen reported in scientific literature (PMID 31200728)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11056893T
SDP
View Documents
Novus Biologicals
NB11056893SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB11056893
SDP
View Documents
Novus Biologicals
NB11056893
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

ATG9A Polyclonal specifically detects ATG9A in Human, Mouse, Rat, Alligator, Bovine, Chicken, Drosophila, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

ATG9A
Western Blot 2 ug/ml, Flow Cytometry, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24705551), Immunoprecipitation reported in scientific literature (PMID 27316455), Immunohistochemistry-Paraffin 1:100-1:250, Immunohistochemistry-Frozen reported in scientific literature (PMID 31200728)
Polyclonal
Rabbit
Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway
PBS with 0.02% Sodium Azide
APG9 autophagy 9-like 1, APG9 autophagy 9-like 1 (S. cerevisiae), APG9L1APG9-like 1, ATG9 autophagy related 9 homolog A (S. cerevisiae), autophagy 9-like 1 protein, autophagy-related protein 9A, FLJ22169, mATG9, MGD3208
ATG9A
IgG
Affinity Purified
95 kDa
1.0 mg/ml
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
RUO
Human, Mouse, Rat, Alligator, Bovine, Chicken, Drosophila, Primate
Q7Z3C6
79065
A synthetic peptide made to a region within the C-terminus of human ATG9A (between residues 750-839). [Swiss-Prot: Q7Z3C6].
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.