Learn More
Invitrogen™ ATG9B Polyclonal Antibody

Description
A suggested positive control is Hela cell lysate. PA5-20998 can be used with blocking peptide PEP-1112.
Specifications
Specifications
| Antigen | ATG9B |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ATG9B |
| Gene Accession No. | Q674R7, Q6EBV9 |
| Gene Alias | Apg912; Apg9l2; APG9-like 2; Apgdc2; ATG9; ATG9 autophagy related 9 homolog B; Atg9b; autophagy 9-like 2 protein; autophagy related 9B; autophagy-related protein 9B; endothelial nitric oxide synthase antisense; eONE; Gm574; nitric oxide synthase 3 antisense; nitric oxide synthase 3-overlapping antisense gene protein; NOS3AS; Protein sONE; RGD1560887; SONE |
| Gene Symbols | ATG9B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.