Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATG9B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577776
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577776 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577776 Supplier Invitrogen™ Supplier No. PA577776
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A 1:1,000 dilution of PA5-77776 was sufficient for detection of ATG9B in 15 μg of Human HeLa Cell lysates by ECL immunoblot analysis using goat anti-rabbit IgG:HRP as the secondary antibody. Detects approximately 100 kDa.

Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. ATG9B plays a role in autophagy and it's highly expressed in placenta and pituitary gland.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG9B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene ATG9B
Gene Accession No. Q674R7, Q6EBV9
Gene Alias Apg912; Apg9l2; APG9-like 2; Apgdc2; ATG9; ATG9 autophagy related 9 homolog B; Atg9b; autophagy 9-like 2 protein; autophagy related 9B; autophagy-related protein 9B; endothelial nitric oxide synthase antisense; eONE; Gm574; nitric oxide synthase 3 antisense; nitric oxide synthase 3-overlapping antisense gene protein; NOS3AS; Protein sONE; RGD1560887; SONE
Gene Symbols ATG9B
Host Species Rabbit
Immunogen Synthetic peptide from the N-terminal of Human ATG9B (aa 110-121).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213948, 285973
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.