Learn More
Invitrogen™ ATG9B Polyclonal Antibody

Description
Immunogen sequence: HPLWRPPGHS SKFLGHLWGR VQQDAAAWGA TSARGPSTPG VLSNCTSPLP EAFLANLFVH PLLPPRDLSP TAPCPAAATA SLLASISRIA QDPSSVSPGG TGGQKLAQLP ELASAEMSLH VIYLHQLHQQ QQQQEPWGEA AASILSRPCS SPSQPPSPDE EKPSWSSDGS SPASSPRQQW GTQKARNLFP GGFQVTTDTQ KEPDRASCTD; Positive Samples: HepG2, HL-60, Mouse liver, Mouse brain, Rat brain; Cellular Location: Cytoplasmic vesicle, Multi-pass membrane protein, autophagosome membrane.
Specifications
Specifications
| Antigen | ATG9B |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.42 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ATG9B |
| Gene Accession No. | Q674R7, Q6EBV9 |
| Gene Alias | Apg912; Apg9l2; APG9-like 2; Apgdc2; ATG9; ATG9 autophagy related 9 homolog B; Atg9b; autophagy 9-like 2 protein; autophagy related 9B; autophagy-related protein 9B; endothelial nitric oxide synthase antisense; eONE; Gm574; nitric oxide synthase 3 antisense; nitric oxide synthase 3-overlapping antisense gene protein; NOS3AS; Protein sONE; RGD1560887; SONE |
| Gene Symbols | ATG9B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.