Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATGL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110189 Supplier Invitrogen™ Supplier No. PA5110189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GASAGALTAT ALVTGVCLGE AGAKFIEVSK EARKRFLGPL HPSFNLVKII RSFLLKVLPA DSHEHASGRL GISLTRVSDG ENVIISHFNS KDELIQANVC SGFIPVYCGL IPPSLQGVRY VDGGISDNLP LYELKNTITV SPFSGESDIC PQDSSTNIHE LRVTNTSIQF NLRNLYRLSK ALFPPEPLVL REMCKQGYRD GLRFLQRNGL LNR.

Lipolytic enzymes are required for mobilization of fatty acids from triglyceride stores in adipose tissue. Energy homeostasis is affected by dysfunctional lipolysis and may contribute to the pathogenesis of obesity and insulin resistance. Until recently, hormone-sensitive lipase (HSL) was the only enzyme known to hydrolyze triglycerides in mammalian adipose tissue. It is now thought that a second enzyme, adipose triglyceride lipase (ATGL), catalyzes the initial step in triglyceride hydrolysis. ATGL is highly expressed in adipose tissue of mice and humans. It exhibits high substrate specificity for triacylglycerol and is associated with lipid droplets. Inhibition of ATGL markedly decreases total adipose acyl-hydrolase activity. Thus, ATGL and HSL coordinately catabolize stored triglycerides in adipose tissue of mammals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATGL
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PNPLA2
Gene Accession No. P0C548, Q8BJ56, Q96AD5
Gene Alias 0610039C21Rik; 1110001C14Rik; adipose triglyceride lipase; ATGL; ATGL DESNUTRIN; calcium-independent phospholipase A2; Desnutrin; DKFZp667M109; FP17548; IPLA2 zeta; iPLA2zeta; IPLA2-zeta; mutant patatin-like phospholipase domain containing 2; patatin like phospholipase domain containing 2; patatin-like phospholipase domain containing 2; patatin-like phospholipase domain containing protein 2; patatin-like phospholipase domain-containing protein 2; PEDF R; PEDF-R; pigment epithelium-derived factor; plpl; plpl2; Pnpla2; RGD1309044; transport-secretion protein; Transport-secretion protein 2; transport-secretion protein 2.2; triglyceride hydrolase; TTS 2.2; TTS2; TTS2.2; TTS-2.2
Gene Symbols PNPLA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 45-257 of human PNPLA2 (NP_0651091).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361676, 57104, 66853
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.