Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATGL Recombinant Rabbit Monoclonal Antibody (1H6)
GREENER_CHOICE

Catalog No. FEMA550194
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550194 Supplier Invitrogen™ Supplier No. MA550194
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

ATGL Recombinant Monoclonal Antibody for Flow, ELISA

Lipolytic enzymes are required for mobilization of fatty acids from triglyceride stores in adipose tissue. Energy homeostasis is affected by dysfunctional lipolysis and may contribute to the pathogenesis of obesity and insulin resistance. Until recently, hormone-sensitive lipase (HSL) was the only enzyme known to hydrolyze triglycerides in mammalian adipose tissue. It is now thought that a second enzyme, adipose triglyceride lipase (ATGL), catalyzes the initial step in triglyceride hydrolysis. ATGL is highly expressed in adipose tissue of mice and humans. It exhibits high substrate specificity for triacylglycerol and is associated with lipid droplets. Inhibition of ATGL markedly decreases total adipose acyl-hydrolase activity. Thus, ATGL and HSL coordinately catabolize stored triglycerides in adipose tissue of mammals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATGL
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Clone 1H6
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PNPLA2
Gene Accession No. Q96AD5
Gene Alias 0610039C21Rik; 1110001C14Rik; adipose triglyceride lipase; ATGL; ATGL DESNUTRIN; calcium-independent phospholipase A2; Desnutrin; DKFZp667M109; FP17548; IPLA2 zeta; iPLA2zeta; IPLA2-zeta; mutant patatin-like phospholipase domain containing 2; patatin like phospholipase domain containing 2; patatin-like phospholipase domain containing 2; patatin-like phospholipase domain containing protein 2; patatin-like phospholipase domain-containing protein 2; PEDF R; PEDF-R; pigment epithelium-derived factor; plpl; plpl2; Pnpla2; RGD1309044; transport-secretion protein; Transport-secretion protein 2; transport-secretion protein 2.2; triglyceride hydrolase; TTS 2.2; TTS2; TTS2.2; TTS-2.2
Gene Symbols PNPLA2
Host Species Rabbit
Immunogen A synthesized peptide derived from Human PNPLA2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57104
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.