Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Atherin Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204030AF405
Description
Atherin Polyclonal antibody specifically detects Atherin in Human samples. It is validated for Western BlotSpecifications
Atherin | |
Polyclonal | |
Western Blot | |
SAMD1 sterile alpha motif domain containing 1 | |
The immunogen recognized by this antibody maps to this antibody Motif Domain Containing 1 using the numbering given in entry NP_612361.1 (GeneID 90378). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
90378 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction