Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
MP Biomedicals™ Atherogenic Diet
Shop All MP Biomedicals, Inc Products
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                10 kg
                            
                        
                            
                                20 kg
                            
                        
                            
                                50 kg
                            
                        
                    Description
- Atherogenic diet is rich in cholesterol and other atherogenic factors
- Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease
 
            Specifications
Specifications
| Product Type | Animal Diet | 
| For Use With (Application) | Rodent diet | 
| Quantity | 50 kg | 
| Storage Requirements | 2°C to 8°C | 
Safety and Handling
                            
                            WARNING: Reproductive Harm - www.P65Warnings.ca.gov 
                        
                    
                
            
            
            
        Atherogenic diet is rich in cholesterol and other atherogenic factors. Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease.
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            