Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATIC/AICAR transformylase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982328
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2328 Supplier Bioss Supplier No. BS12544R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATIC is a 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase which helps catalyze the penultimate and final steps of the de novo purine biosynthetic pathway. The human ATIC cDNA encodes a deduced 591-amino acid protein that is 81% identical to the chicken sequence.

Specifications

Antigen ATIC/AICAR transformylase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ATIC
Gene Accession No. P31939
Gene Alias 2610509C24Rik; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase L homeolog; 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotide transformylase/inosinicase; AA536954; AICAR; AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme; AICAR transformylase; AICARFT; AICARFT/IMPCHASE; ATIC; atic.L; AW212393; Bifunctional purine biosynthesis protein PURH; Bifunctional purine biosynthesis protein PURH, N-terminally processed; epididymis secretory sperm binding protein Li 70p; HEL-S-70p; IMP cyclohydrolase; IMP synthase; IMPCHASE; Inosinicase; OK/SW-cl.86; Phosphoribosylaminoimidazolecarboxamide formyltransferase; phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase; PURH; XELAEV_18044849mg
Gene Symbols ATIC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ATIC/AICAR transformylase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 471
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.