Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATIC Antibody (OTI1D2), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270220MFV450
Description
ATIC Monoclonal antibody specifically detects ATIC in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow CytometrySpecifications
ATIC | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Flow Cytometry | |
OTI1D2 | |
Flow Cytometry | |
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMPcyclohydrolase, 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotidetransformylase/inosinicase, AICAR, AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme, AICARFT, AICARFT/IMPCHASE, bifunctional purine biosynthesis protein PURH, IMPCHASE, phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase, PURHFLJ93545 | |
Full length human recombinant protein of human ATIC(NP_004035) produced in HEK293T cell. | |
0.1 mL | |
Stem Cell Markers | |
471 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction