Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATIC Monoclonal Antibody (OTI2C10), TrueMAB™, OriGene
SDP

Catalog No. 501675211
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-5211 100 μL
50-254-0255 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-5211 Supplier OriGene Supplier No. TA504554
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATIC is a 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase which helps catalyze the penultimate and final steps of the de novo purine biosynthetic pathway. The human ATIC cDNA encodes a deduced 591-amino acid protein that is 81% identical to the chicken sequence.

Specifications

Antigen ATIC
Applications Western Blot
Classification Monoclonal
Clone OTI2C10
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ATIC
Gene Accession No. O35567, P31939, Q9CWJ9
Gene Alias AICAR, AICARFT, HEL-S-70p, IMPCHASE, PURH
Gene Symbols ATIC
Host Species Mouse
Immunogen Full length human recombit protein of human ATIC produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108147, 471, 488513, 81643
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.