Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATIC Monoclonal Antibody (OTI2C10), TrueMAB™, OriGene
SDP

Catalog No. 502540255
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0255 30 μL
50-167-5211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0255 Supplier OriGene Supplier No. TA504554S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATIC Monoclonal antibody specifically detects ATIC in Canine, Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen ATIC
Applications Western Blot
Classification Monoclonal
Clone OTI2C10
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ATIC
Gene Accession No. O35567, P31939, Q9CWJ9
Gene Alias 153761_at; 2610509C24Rik; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase L homeolog; 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotide transformylase/inosinicase; AA536954; AICAR; AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme; AICAR transformylase; AICARFT; AICARFT/IMPCHASE; atic; atic.L; AW212393; BEST:GH11240; Bifunctional purine biosynthesis protein PURH; Bifunctional purine biosynthesis protein PURH, N-terminally processed; CG11089; CG11089-PA; CG11089-PB; CT30991; Dmel\CG11089; Dmel_CG11089; epididymis secretory sperm binding protein Li 70p; HEL-S-70p; IMP cyclohydrolase; IMP synthase; IMPCHASE; Inosinicase; OK/SW-cl.86; Phosphoribosylaminoimidazolecarboxamide formyltransferase; phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase; PURH; RE29555p; uncharacterized protein; XELAEV_18044849mg
Host Species Mouse
Immunogen Full length human recombinant protein of human ATIC produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108147, 471, 488513, 81643
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.