Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATIC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117021 Supplier Invitrogen™ Supplier No. PA5117021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, SW480, NCI-H460, HepG2, SW620, HT-29, K-562 Immunogen sequence: MAPGQLALFS VSDKTGLVEF ARNLTALGLN LVASGGTAKA LRDAGLAVRD VSELTGFPEM LGGRVKTLHP AVHAGILARN IPEDNADMAR LDFNLIRVVA CNLYPFVKTV ASPGVTVEEA VEQIDIGGVT LLRAAAKNHA RVTVVCEPED YVVVSTEMQS SESKDTSLET RRQLALKAFT HTAQYDEAIS DYFRKQYSKG VSQMPLRYGM NPHQTPAQLY TLQPKLPITV LNGAPGFINL CDALNAWQLV KELKEALGIP AAASFKHVSP AGAAVGIPLS EDEAKVCMVY DLYKTLTPIS.

ATIC is a 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase which helps catalyze the penultimate and final steps of the de novo purine biosynthetic pathway. The human ATIC cDNA encodes a deduced 591-amino acid protein that is 81% identical to the chicken sequence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATIC
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATIC
Gene Accession No. P31939
Gene Alias 153761_at; 2610509C24Rik; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase L homeolog; 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotide transformylase/inosinicase; AA536954; AICAR; AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme; AICAR transformylase; AICARFT; AICARFT/IMPCHASE; atic; atic.L; AW212393; BEST:GH11240; Bifunctional purine biosynthesis protein PURH; Bifunctional purine biosynthesis protein PURH, N-terminally processed; CG11089; CG11089-PA; CG11089-PB; CT30991; Dmel\CG11089; Dmel_CG11089; epididymis secretory sperm binding protein Li 70p; HEL-S-70p; IMP cyclohydrolase; IMP synthase; IMPCHASE; Inosinicase; OK/SW-cl.86; Phosphoribosylaminoimidazolecarboxamide formyltransferase; phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase; PURH; RE29555p; uncharacterized protein; XELAEV_18044849mg
Gene Symbols ATIC
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ATIC (NP_0040352).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 471
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.