Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569621 Supplier Invitrogen™ Supplier No. PA569621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 75%.

ATL2 gene ontology annotations related to this gene include GTPase activity; GTP binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ATL2
Gene Accession No. Q8NHH9
Gene Alias 2010110I21Rik; AA407293; ADAMTSL-2; ADP-ribosylation factor-like 6 interacting protein 2; ADP-ribosylation factor-like protein 6-interacting protein 2; ADP-ribosylation-like factor 6 interacting protein 2; aip-2; ARL3IP2; ARL-6 interacting protein-2; ARL-6-interacting protein 2; ARL6IP2; Atl2; atlastin GTPase 2; atlastin GTPase 2; atlastin-2; atlastin2; atlastin-2; AV334690; fc06e01; GPHYSD1; wu:fc06e01; wu:fc06e01 protein; wu:fv60d03
Gene Symbols ATL2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human ATL2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64225
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.