Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATL2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ATL2 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1115
![]() ![]() |
Proteintech Group Inc
166881AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ATL2 gene ontology annotations related to this gene include GTPase activity; GTP binding.Specifications
ATL2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8NHH9 | |
64225 | |
ATL2 Fusion Protein Ag10030 | |
Primary | |
-20°C | |
ATL2 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Aip 2, ARL 6 interacting protein 2, ARL3IP2, ARL6IP2, ATL2, Atlastin 2, atlastin GTPase 2, atlastin2 | |
ATL2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title