Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598495
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598495 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598495 Supplier Invitrogen™ Supplier No. PA598495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATL3 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

ATL3 is GTPase tethering membranes through formation of trans-homooligomer and mediating homotypic fusion of endoplasmic reticulum membranes. ATL3 is function in endoplasmic reticulum tubular network biogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATL3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Atl3
Gene Accession No. Q6DD88
Gene Alias 4633402C03Rik; 5730596K20Rik; AI465397; Atl3; atlastin 3; atlastin GTPase 3; Atlastin-3; AW228836; HSN1F; I79_021367; RGD1309871
Gene Symbols Atl3
Host Species Rabbit
Immunogen Recombinant Human Atlastin-3 protein (1-187AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25923
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.