Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113835 Supplier Invitrogen™ Supplier No. PA5113835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DHFKKTKKMG GKDFSFRYQQ ELEEEIKELY ENFCKHNGSK NVFSTFRTPA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

ATL3 is GTPase tethering membranes through formation of trans-homooligomer and mediating homotypic fusion of endoplasmic reticulum membranes. ATL3 is function in endoplasmic reticulum tubular network biogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATL3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Atl3
Gene Accession No. Q6DD88
Gene Alias 4633402C03Rik; 5730596K20Rik; AI465397; Atl3; atlastin 3; atlastin GTPase 3; Atlastin-3; AW228836; HSN1F; I79_021367; RGD1309871
Gene Symbols Atl3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DKFZP564J0863.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25923
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.