Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 317 Hi-Vis Latex Coated Seamless Knit Polyester Gloves

Catalog No. 19020331 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-020-331 Large
19-020-332 X-Large
19-020-333 Small
19-020-330 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-020-331 Supplier SHOWA™ Supplier No. 317L09
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves are designed to provide users with increased protection in low-light applications, with a textured latex coating for enhanced grip capabilities.

  • Hardwearing gloves designed to provide protection in low-light, ideal for use indoors and out
  • Textured latex coating on the palms protects hands and enhances grip
  • Elastic wrist cuff keeps dirt and debris out of the gloves

Specifications

Construction Seamless Knit
Coating Material Latex
Color Hi-vis Yellow, Orange
Exterior Finish Fully Textured
Hand Specific
Material Polyester
Product Type Seamless Knit Gloves
Size Large
EN388 Standard 2142X
For Use With (Application) Heavy Equipment Handling, Maintenance, and Shipping/Receiving
Glove Cuff Style Knit
Series 317
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.