Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Atlastin-1, Mouse, Unlabeled, Clone: 3194,

Catalog No. MABN1831MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1831MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1831MI Supplier MilliporeSigma™ Supplier No. MABN1831
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Atlastin-1 (UniProt Q8WXF7; also known as Brain-specific GTP-binding protein, GBP-3, GTP-binding protein 3, Guanine nucleotide-binding protein 3, hGBP3, Spastic paraplegia 3 protein A) is encoded by the ATL1 (also known as GBP3, HSN1D, SPG3, SPG3A) gene (Gene ID 51062) in human. Atlastin-1 is a dynamin/Mx/guanylate-binding protein superfamily member with three conserved guanylate-binding/GTPase active site motifs, P-loop (a.a. 74-81), DxxG (a.a. 146-149), and RD (217-218). Atlastin-1 is a multimeric integral membrane protein localized on ER and cis-Golgi apparatus and widely expressed in many tissues, including the smooth muscle, adrenal gland, kidney, testis, lung, and brain. Atlastin-1 interacts with the microtubule-severing ATPase spastin as well as with the DP1/Yop1p and reticulon families of ER-shaping proteins, Atlastin-1 knockdown results in decreased number of neuronal processes and impaired axon formation in cultured cortical neurons, while atlastin-1 overexpression increases total dendrite length both in vivo and in vitro. ALT1 gene mutations cause abnormal ER morphology and are linked to hereditary spastic paraplegias (HSPs), a genetically heterogeneous group of neurological disorders, includiing SPG3 (spastic paraplegia 3, autosomal dominan) and HSN1D (neuropathy, hereditary sensory, 1D), characterized by lower limb spasticity and weakness. Human atlastin-1 is a 492-amino acid protein that passes through the ER membrane twice (a.a. 450-470, 472-492), having both its N- and C-terminal ends at the cytoplasmic side (a.a. 1-449, 493-558).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Atlastin-1
Applications Western Blot
Classification Monoclonal
Clone 3194
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene ATL1, GBP3, HSN1D, SPG3, SPG3A
Gene Accession No. Q8WXF7
Host Species Mouse
Immunogen Linear peptide corresponding to the N-terminal sequence of human Atlastin-1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_056999
Test Specificity Clone 3194 targets an N-terminal epitope present in both spliced isoforms of human Atlastin-1 reported by UniProt (Q8WXF7).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.